Digi-Sense J20-1-304 Thermocouple, Wire, Type-J, 20-Gauge, Glass Braid, 1000-ft Spool
Representative image only.
Digi-Sense J20-1-304 Thermocouple, Wire, Type-J, 20-Gauge, Glass Braid, 1000-ft Spool
Digi-Sense | Item: 0854217
Make your own extension cables
₹150,000.00INR / Each
₹150,000.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeJ
  • MaterialFiberglass
  • Wire Gauge (Ga)20
  • Error Limit±4°F (±2.2°C) from 32 to 545°F (0 to 285°C); ±0.75% reading from 545 to 1400°F (285 to 750°C)
  • Inner InsulationFiberglass Braid
  • Outer InsulationFiberglass Braid
  • Stranded / SolidSolid
  • OverbraidNo
  • Min Temperature (° F)-100
  • Max Temperature (° F)900
  • Min Temperature (° C)-73
  • Max Temperature (° C)482
  • Length (ft)1000
  • Size (in)0.060 in x 0.106 in
  • DescriptionThermocouple, Wire, Type-J, 20-Gauge, Glass Braid, 1000-ft Spool