Digi-Sense K20-1-507 Thermocouple Wire Type K 20-Gauge, FEP Insulation, 100-ft Spool
Representative image only.
Digi-Sense K20-1-507 Thermocouple Wire Type K 20-Gauge, FEP Insulation, 100-ft Spool
Digi-Sense | Item: 0854121
Make your own extension cables
₹33,100.00INR / Each
₹33,100.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeK
  • MaterialFluorinated ethylene propylene (FEP)
  • Wire Gauge (Ga)20
  • Error Limit±2% reading from -330 to -165°F (-200 to -110°C); ±4°F (±2.2°C) from -165 to 545°F (-110 to 285°C); ±0.75% reading from 545 to 2300°F (285 to 1250°C)
  • Inner InsulationFluorinated ethylene propylene (FEP)
  • Outer InsulationFluorinated ethylene propylene (FEP)
  • Stranded / SolidSolid
  • OverbraidNo
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)100
  • Size (in)0.068 in x 0.116 in
  • DescriptionThermocouple Wire Type K 20-Gauge, FEP Insulation, 100-ft Spool
  • Warranty10 month