Digi-Sense Thermocouple Probe Wire, Type J, 20-Gauge, Glass Braid with SS Overbraid, 100ft
Representative image only.
Digi-Sense Thermocouple Probe Wire, Type J, 20-Gauge, Glass Braid with SS Overbraid, 100ft
Digi-Sense | Item: 0854138
Make your own extension cables
₹38,200.00INR / Each
₹38,200.00INR / Each
Product Description
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
Specifications
  • Connector TypeJ
  • MaterialFiberglass
  • Wire Gauge (Ga)20
  • Inner InsulationFiberglass Braid
  • Outer InsulationFiberglass Braid
  • Stranded / SolidSolid
  • OverbraidYes
  • Special InformationStainless steel overbraid
  • Min Temperature (° F)-100
  • Max Temperature (° F)900
  • Min Temperature (° C)-73
  • Max Temperature (° C)482
  • Length (ft)100
  • DescriptionThermocouple Probe Wire, Type J, 20-Gauge, Glass Braid with SS Overbraid, 100-ft Spool