Cole-Parmer® 96-Well Deep Well Plates
Excellent for assays,sample storage and mixing
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Notched corners to facilitate orientation
  • Compatible with most automated liquid handling systems
Deep-well plates made of polypropylene (PP) can be autoclaved at 121°C for 20 minutes. Raised rims provide efficient sealing to minimize the risk of cross-contamination. Round U-shaped wells reduce sample loss while pipetting. Available gamma sterilized.
Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination.
17,600.00 - ₹92,400.00INR / Pkg of 96
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Well Volume (mL) Description
Availability Pricing
COMPARE
1.1 Deep Well Plate, 1.1 mL, Round Wells, U Shaped, non sterile; 96/Pk
₹28,500.00INR / Pkg of 96
COMPARE
1.1 Deep Well Plate, 1.1mL, Round Wells, U-shaped, sterile; 96/Pk
₹29,700.00INR / Pkg of 96
COMPARE
2.2 Deep Well Plate, 2.2 mL, Round Wells, V-Bottom, non sterile; 48/Pk
₹17,600.00INR / Pkg of 48
COMPARE
2.2 Deep Well Plate, 2.2 mL, Round Wells, V-Bottom, Sterile; 48/Pk
₹17,900.00INR / Pkg of 48
COMPARE
1.6 96-Well Deep Well Plate, 1.6 mL, U Type, Non-Sterile; 50/Cs
₹30,700.00INR / Case of 50
COMPARE
1.6 96-Well Deep Well Plate, 1.6 mL, U Type, Sterile; 50/Cs
₹33,200.00INR / Case of 50
COMPARE
2.2 96-Well Deep Well Plate, 2.2 mL, U Type, Non-Sterile; 50/Cs
₹21,200.00INR / Case of 50
COMPARE
2.2 96-Well Deep Well Plate, 2.2 mL, U Type, Sterile; 50/Cs
₹24,500.00INR / Case of 50
COMPARE
Sealing Film for 96-Well Deep Well Plate, Non-Sterile, 1000/Cs
₹75,300.00INR / Case of 1000
COMPARE
Sealing Film for 96-Well Deep Well Plate, Sterile, 1000/Cs
₹92,400.00INR / Case of 1000
COMPARE
Sealing Pad for 96-Well Deep Well Plate, Non-Sterile, 100/Cs
₹21,500.00INR / Case of 100
COMPARE
Sealing Pad for 96-Well Deep Well Plate, Sterile, 100/Cs
₹22,300.00INR / Case of 100
Showing 1- 12 of 12