Cole-Parmer 96-Well Deep Well Plate, 2.2 mL, U Type, Sterile; 50/Cs

Representative image only.

Cole-Parmer 96-Well Deep Well Plate, 2.2 mL, U Type, Sterile; 50/Cs

Cole-Parmer | Item: 1634493

Excellent for assays,sample storage and mixing

₹24,500.00INR / Case of 50

₹24,500.00INR / Case of 50
Product Description

Key Features

  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Notched corners to facilitate orientation
  • Compatible with most automated liquid handling systems

More About this Item

Deep-well plates made of polypropylene (PP) can be autoclaved at 121°C for 20 minutes. Raised rims provide efficient sealing to minimize the risk of cross-contamination. Round U-shaped wells reduce sample loss while pipetting. Available gamma sterilized.

Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination.

Sterile
Specifications
  • Number of Wells96
  • Well Volume (mL)2.2
  • Well Volume (µL)2200
  • Well ShapeU Type
  • Lid (Included)No
  • SterileSterile
  • Description96-Well Deep Well Plate, 2.2 mL, U Type, Sterile; 50/Cs
Reviews
Q&A