Cole-Parmer Sealing Film for 96-Well Deep Well Plate, Non-Sterile, 1000/Cs
Representative image only.
Cole-Parmer Sealing Film for 96-Well Deep Well Plate, Non-Sterile, 1000/Cs
Cole-Parmer | Item: 1634494
Excellent for assays,sample storage and mixing
₹75,300.00INR / Case of 1000
₹75,300.00INR / Case of 1000
Product Description
Key Features
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Notched corners to facilitate orientation
  • Compatible with most automated liquid handling systems
More About this Item
Deep-well plates made of polypropylene (PP) can be autoclaved at 121°C for 20 minutes. Raised rims provide efficient sealing to minimize the risk of cross-contamination. Round U-shaped wells reduce sample loss while pipetting. Available gamma sterilized.
Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination.
Specifications
  • SterileNon-Sterile
  • DescriptionSealing Film for 96-Well Deep Well Plate, Non-Sterile, 1000/Cs