Cole-Parmer Deep Well Plate, 2.2 mL, Round Wells, V-Bottom, Sterile; 48/PK

Representative image only.

Cole-Parmer Deep Well Plate, 2.2 mL, Round Wells, V-Bottom, Sterile; 48/PK

Cole-Parmer | Item: 1597805

Excellent for assays,sample storage and mixing

₹17,900.00INR / Pkg of 48

₹17,900.00INR / Pkg of 48
Product Description

Key Features

  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Notched corners to facilitate orientation
  • Compatible with most automated liquid handling systems
  • 1 year warranty

More About this Item

Deep-well plates made of polypropylene (PP) can be autoclaved at 121°C for 20 minutes. Raised rims provide efficient sealing to minimize the risk of cross-contamination. Round U-shaped wells reduce sample loss while pipetting. Available gamma sterilized.

Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination.

1 Year Warranty
Specifications
  • Well Volume (mL)2.2
  • Well Volume (µL)2200
  • Max Working Well Volume (mL)2.2
  • Well ShapeV Bottom
  • SterileNon-Sterile
  • MaterialPolypropylene
  • DescriptionDeep Well Plate, 2.2 mL, Round Wells, V-Bottom, Sterile; 48/Pk
  • Warranty1 year
Reviews
Q&A