Cole-Parmer 96-Well Deep Well Plate, 1.6 mL, U Type, Non-Sterile; 50/Cs
Representative image only.
Cole-Parmer 96-Well Deep Well Plate, 1.6 mL, U Type, Non-Sterile; 50/Cs
Cole-Parmer | Item: 1634490
Excellent for assays,sample storage and mixing
₹30,700.00INR / Case of 50
₹30,700.00INR / Case of 50
Product Description
Key Features
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Notched corners to facilitate orientation
  • Compatible with most automated liquid handling systems
More About this Item
Deep-well plates made of polypropylene (PP) can be autoclaved at 121°C for 20 minutes. Raised rims provide efficient sealing to minimize the risk of cross-contamination. Round U-shaped wells reduce sample loss while pipetting. Available gamma sterilized.
Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination.
Specifications
  • Number of Wells96
  • Well Volume (mL)1.6
  • Well Volume (µL)1600
  • Well ShapeU Type
  • Lid (Included)No
  • SterileNon-Sterile
  • Description96-Well Deep Well Plate, 1.6 mL, U Type, Non-Sterile; 50/Cs