Cole-Parmer Sealing Pad for 96-Well Deep Well Plate, Sterile, 100/Cs
Representative image only.
Cole-Parmer Sealing Pad for 96-Well Deep Well Plate, Sterile, 100/Cs
Cole-Parmer | Item: 1634497
Excellent for assays,sample storage and mixing
₹22,300.00INR / Case of 100
₹22,300.00INR / Case of 100
Product Description
Key Features
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Notched corners to facilitate orientation
  • Compatible with most automated liquid handling systems
More About this Item
Deep-well plates made of polypropylene (PP) can be autoclaved at 121°C for 20 minutes. Raised rims provide efficient sealing to minimize the risk of cross-contamination. Round U-shaped wells reduce sample loss while pipetting. Available gamma sterilized.
Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination.
Sterile
Specifications
  • SterileSterile
  • DescriptionSealing Pad for 96-Well Deep Well Plate, Sterile, 100/Cs