TSI 9565-X VelociCalc Ventilation Meter Only
Representative image only.
TSI 9565-X VelociCalc Ventilation Meter Only
TSI | Mfr: 9565-X| Item: 1030248
TSI's most advanced thermoanemometers
₹466,000.00INR / Each
for an additional
₹47,800.00INR / Each
for an additional
₹60,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹466,000.00INR / Each
Buy It Together
Current Item
TSI 9565-X VelociCalc Ventilation Meter Only
TSI 9565-X VelociCalc Ventilation Meter Only
1030248
₹466,000.00INR / Each
+ +
InnoCal NIST-Traceable Calibration; Anemometer, Vane-Type
1710101
₹47,800.00INR / Each
=
Total Price 768800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
4 AA batteries.
Key Features
  • Bluetooth® printer capability
  • Manual or continuous data logging
  • Displays up to 5 measurements simultaneously
  • Log up to 26,500 samples and 100 test IDs
  • Comes with NIST-traceable calibration report supplied by manufacturer
More About this Item
The TSI series of portable, handheld, multi-function ventilation test instruments — available with or without a differential pressure sensor — is designed to work with a wide range of plug-in probes. The probes allow you to make various measurements by simply plugging in a different probe that has the features and functions best suited for a particular application. They are designed to measure air velocity, temperature, humidity, CO and CO2. Probes feature a "SMART" plug-in connection that stores all the calibration data, so a new probe can be ordered at any time without the need of recalibrating the meter.
The meter can perform calculations for air flow, heat flow, turbulence, wet bulb and dew point temperature — then data can be downloaded to a PC or portable printer for analysis. Statistics function displays number of recorded samples, average, and Min/Max. Display automatically stabilizes while measuring in fluctuating flow applications. Backlit display allows easy viewing in low-lit areas.
Multi-Function Ventilation Meter with Dataloging (No probe included)
Conformite EuropeenneTwo year warranty
Specifications
  • Number Of Temperature Channels1
  • Min Temperature (° F)40
  • Max Temperature (° F)113
  • Min Temperature (° C)5
  • Max Temperature (° C)45
  • Data LoggingYes
  • Data Storage (samples)26,500 samples and 100 test IDs
  • Logging Intervals1 second to 1 hour
  • Width (in)4
  • Height (in)2
  • Length (in)8 3/8
  • Battery4 AA (included)
  • DescriptionVelociCalc Ventilation Meter Only