TSI 982 VelociCalc IAQ Probe, CO2, Temp, Humidity, CO
Representative image only.
TSI 982 VelociCalc IAQ Probe, CO2, Temp, Humidity, CO
TSI | Mfr: 982| Item: 1037239
₹670,000.00INR / Each
₹670,000.00INR / Each
Buy It Together
Current Item
TSI 982 VelociCalc IAQ Probe, CO2, Temp, Humidity, CO
TSI 982 VelociCalc IAQ Probe, CO2, Temp, Humidity, CO
1037239
₹670,000.00INR / Each
+
TSI 9565-X VelociCalc Ventilation Meter Only
1030248
₹466,000.00INR / Each
=
Total Price 1136000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Keep your TSI ventilation meter working dependably with these parts and accessories.
NIST Traceable CalibrationTwo year warranty
Specifications
  • Min Temperature (° F)14
  • Max Temperature (° F)140
  • Min Temperature (° C)-10
  • Max Temperature (° C)60
  • Temperature Accuracy±0.5°F (±0.3°C)
  • Temperature Resolution0.1°F (0.1°C)
  • RH Resolution0.1% RH
  • RH Accuracy±3%
  • DescriptionVelociCalc IAQ Probe, CO2, Temp, Humidity, CO