TSI VelociCalc Multi-Function Ventilation Meters
TSI's most advanced thermoanemometers
  • Bluetooth® printer capability
  • Manual or continuous data logging
  • Displays up to 5 measurements simultaneously
  • Log up to 26,500 samples and 100 test IDs
  • Comes with NIST-traceable calibration report supplied by manufacturer
The TSI series of portable, handheld, multi-function ventilation test instruments — available with or without a differential pressure sensor — is designed to work with a wide range of plug-in probes. The probes allow you to make various measurements by simply plugging in a different probe that has the features and functions best suited for a particular application. They are designed to measure air velocity, temperature, humidity, CO and CO2. Probes feature a "SMART" plug-in connection that stores all the calibration data, so a new probe can be ordered at any time without the need of recalibrating the meter.
The meter can perform calculations for air flow, heat flow, turbulence, wet bulb and dew point temperature — then data can be downloaded to a PC or portable printer for analysis. Statistics function displays number of recorded samples, average, and Min/Max. Display automatically stabilizes while measuring in fluctuating flow applications. Backlit display allows easy viewing in low-lit areas.
466,000.00 - ₹957,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
TSI 9565-X VelociCalc Ventilation Meter Only 1030248 Mfr # 9565-X
COMPARE
VelociCalc Ventilation Meter Only
₹466,000.00INR / Each
TSI 9565 VelociCalc Ventilation Meter; comes w/Straight Probe 10372-35 1037242 Mfr # 9565
COMPARE
VelociCalc Ventilation Meter; comes w/Straight Probe 10372-35
₹890,000.00INR / Each
TSI 9565-A VelociCalc Ventilation Meter; comes w/Articulating Probe 10372-36 1037243 Mfr # 9565-A
COMPARE
VelociCalc Ventilation Meter; comes w/Articulating Probe 10372-36
₹957,000.00INR / Each
TSI 9565-P VelociCalc Ventilation Meter with differential pressure sensor 1037244 Mfr # 9565-P
COMPARE
VelociCalc Ventilation Meter with differential pressure sensor
₹786,000.00INR / Each