Sartorius Arium® Mini Plus Starter Set with UV Lamp

Representative image only.

Sartorius Arium® Mini Plus Starter Set with UV Lamp

Sartorius | Mfr: H2O-MA-UV-SET-US| Item: 9929445

Starter set includes all the required components to get you up and running

Call For Price

Call For Price

Buy It Together

Current Item

Sartorius Arium® Mini Plus Starter Set with UV Lamp
Sartorius Arium® Mini Plus Starter Set with UV Lamp
9929445

Call For Price
+
Sartorius H2O-CBS-5-S Arium® Mini Storage Bag, 5 L
9929333

₹15,400.00INR / Each
+ =
Total Price 15400.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Arium Mini Plus system with UV lamp, Scientific kit, 5 L bag, and SterilePlus filter.

Key Features

  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Large touch display with self-explanatory navigation
  • Take up a small footprint—only measures 28 cm in width
  • Volume-controlled dispensing in 50 mL steps
  • Delivers up to 1 L/minute
  • Produces ASTM Type I ultrapure water and Type II pure water
  • Stores water in 5 L Bagtank bag
  • UV lamp effectively destroys organic compounds
  • 1 year warranty

More About this Item

Using high-quality water ensures the reproducibility of your results and prevents time-consuming repeat analysis. These compact Arium Mini systems only measure 28 cm (11") wide to easily fit on a benchtop. A high-resolution, touch-activated color display featuring self-explanatory icons is easy to navigate even while wearing gloves. Store favorite functions such repeated dispensing of identical volumes to help increase efficiency while minimizing errors.

Select from Arium Mini, Mini Plus, and Mini Essential depending on your application. All models produce up to 10 L of ASTM Type I ultrapure water per day. Arium Mini and Mini Plus use a closed Bagtank system while Mini Essential has no storage system. For analytical applications, order the Arium Mini with UV lamp to reduce total organic compound (TOC) to = 5 ppb.

Connect Arium Mini Plus water purification system directly to tap water to produce ASTM Type I ultrapure water and Type III pure water. Type I ultrapure water is useful in buffer, media, and reagent preparation and histology, ELISA, AAS, IC and photometry applications. Type III pure water is ideal for general laboratory applications such as rinsing glass and laboratory containers, feed water for laboratory equipment, or feed water for ultrapure water systems.

Arium Mini Plus uses a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals. The integrated UV lamp (185/254 nm) helps reduce total organic compound (TOC) to =5 ppb, making it ideal for analytical applications such as SPE, ICP-MS, GS-MS, LC-MS, and HPLC.

Conformite EuropeenneCanadian Standards Association
Specifications
  • ASTM Water GradeType I & Type III
  • Capacity5 L
  • Water Purification MethodAdsorption by spherical activated carbon, catalyst, reverse osmosis, ion exchange, optional UV irradiation, and by point-of-use particle-removing filtration
  • Max Flow Rate (L/min)1
  • Inlet Port0.25" OD tubing
  • With UV LampYes
  • SourceDirect conection to portable water supply
  • Type I Water (Resistivity)18.2 MΩ × cm
  • Feed Water Requirements (Conductivity)< 1500 µS/cm
  • Type I Water (TOC)≤ 5 ppb
  • Feed Water Requirements (TOC)< 2000 ppb
  • Feed Water Requirements (turbidity)< 1 NTU
  • Type I Water (Conductivity)0.055 μS/cm
  • Type I Water (Bacteria)< 0.01 CFU/mL
  • Feed Water Requirements (pH range)4 to 10
  • Type I Water (Endotoxins)< 0.001 EU/mL
  • Feed Water Requirements (pressure)0.5 to 6 bar
  • Feed Water Requirements (Temperature)2 to 30°C
  • Type III Water (Bacteria)< 0.01 CFU/mL
  • Power (VAC)100 to 240
  • Height (in)20
  • Depth (in)20.9
  • Width (in)11
  • Height (cm)50.9
  • Depth (cm)53.1
  • Width (cm)28
  • DescriptionArium® Mini Plus Starter Set with UV Lamp
  • Warranty2 years
Reviews
Q&A