Idex Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA
Representative image only.
Idex Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA
Idex | Mfr: 7725i| Item: 0201314
Introduce a sample of a solution into a flowing stream of liquid
₹416,000.00INR / Each
₹416,000.00INR / Each
Buy It Together
Current Item
Idex Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA
Idex Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA
0201314
₹416,000.00INR / Each
+
Idex RheBuild® Kit for Valve Model 7725/7725i; 1/EA
0201313
₹61,000.00INR / Each
=
Total Price 477000.00 ADD 2 ITEMS TO CART
Product Description
Includes
20 µL sample loop
Key Features
  • 2-position, 6-port configuration
  • Front loading with switch
  • SS/ceramic/polyimide wetted materials
  • 20 µL sample loading volume
  • 10-32 coned ports for 1/16" OD tubing
  • 1 month warranty
More About this Item
Manual injection valves are ideal for applications that require operator control or injection of smaller sample volumes. Injection valves are a type of switching valve usually featuring an attached sample loop (a length of tubing and fittings that match the angle of the valve ports).
Select from single- or dual-mode valves. Single-mode injection valves are configured for rear loading and requires complete filling of the sample loop. Dual-mode injection valves are configured for front loading and allows either partial or complete filling of the loop. Dual-mode valve introduces sample by syringe through the needle port built into the valve shaft. Valves with MBB design creates a continuous flow between the load and inject positions to eliminate pressure transient shock to the system.
1 Month Warranty
Specifications
  • Outlet Port10-32 coned
  • Inlet Port10-32 coned
  • Max Pressure (PSI)5000
  • Max Pressure (bar)345
  • Wetted MaterialsSS/Ceramic/Polyimide
  • Tubing OD (in)0.0625
  • DescriptionManual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA