Idex® Manual Chromatography Injection Valves
Introduce a sample of a solution into a flowing stream of liquid
  • 2-position, 6-port configuration
Manual injection valves are ideal for applications that require operator control or injection of smaller sample volumes. Injection valves are a type of switching valve usually featuring an attached sample loop (a length of tubing and fittings that match the angle of the valve ports).
Select from single- or dual-mode valves. Single-mode injection valves are configured for rear loading and requires complete filling of the sample loop. Dual-mode injection valves are configured for front loading and allows either partial or complete filling of the loop. Dual-mode valve introduces sample by syringe through the needle port built into the valve shaft. Valves with MBB design creates a continuous flow between the load and inject positions to eliminate pressure transient shock to the system.
134,000.00 - ₹621,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description Tubing OD (in) Wetted Materials
Availability Pricing
Idex Manual Injection Valve, Analytical, Dual Mode, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA 0201311 Mfr # 7725
COMPARE
Manual Injection Valve, Analytical, Dual Mode, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA 0.0625 SS/Ceramic/Polyimide
₹399,000.00INR / Each
Idex Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA 0201314 Mfr # 7725i
COMPARE
Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/Polyimide, 1/16" OD Tubing; 1/EA 0.0625 SS/Ceramic/Polyimide
₹416,000.00INR / Each
Idex Manual Injection Valve, Analytical, Dual Mode with Switch, PEEK/Ceramic/ETFE; 1/EA 0201326 Mfr # 9725i
COMPARE
Manual Injection Valve, Analytical, Dual Mode with Switch, PEEK/Ceramic/ETFE; 1/EA 0.0625 PEEK/Ceramic/PTFE
₹411,000.00INR / Each
Idex Manual Injection Valve, Black, Bulkhead, Polyimide/PTFE, 1/16" OD Tubing; 1/EA 0201492 Mfr # V-451
COMPARE
Manual Injection Valve, Black, Bulkhead, Polyimide/PTFE, 1/16" OD Tubing; 1/EA 0.0625 Polyimide/PTFE
₹134,000.00INR / Each
Idex Manual Injection Valve, Single Mode, SS/Vespel®, 1/16" OD Tubing; 1/EA 0201522 Mfr # 7010
COMPARE
Manual Injection Valve, Single Mode, SS/Vespel®, 1/16" OD Tubing; 1/EA 0.0625 SS/Vespel
₹285,000.00INR / Each
Idex Manual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA 0201525 Mfr # 8125
COMPARE
Manual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA 0.0625 SS/Ceramic/Vespel
₹574,000.00INR / Each
Idex Manual Injection Valve, Analytical, Dual Mode, PEEK/Ceramic/PTFE, 1/16" OD Tubing; 1/EA 0201527 Mfr # 9725
COMPARE
Manual Injection Valve, Analytical, Dual Mode, PEEK/Ceramic/PTFE, 1/16" OD Tubing; 1/EA 0.0625 PEEK/Ceramic/PTFE
₹410,000.00INR / Each
Idex Manual Injection Valve, Preparative, Dual Mode with Switch, PEEK, 1/8" OD Tubing; 1/EA 0201535 Mfr # 3725I
COMPARE
Manual Injection Valve, Preparative, Dual Mode with Switch, PEEK, 1/8" OD Tubing; 1/EA 0.125 PEEK
₹573,000.00INR / Each
Idex Manual Injection Valve, Preparative, Dual Mode with Switch, SS/PEEK, 1/8" OD Tubing; 1/EA 0201536 Mfr # 3725I-038
COMPARE
Manual Injection Valve, Preparative, Dual Mode with Switch, SS/PEEK, 1/8" OD Tubing; 1/EA 0.125 SS/PEEK
₹621,000.00INR / Each
Idex Manual Injection Valve, Preparative, Dual Mode with Switch, No Sample Loop, SS, 1/8" OD Tubing; 1/EA 0201537 Mfr # 3725I-119
COMPARE
Manual Injection Valve, Preparative, Dual Mode with Switch, No Sample Loop, SS, 1/8" OD Tubing; 1/EA 0.125 SS
₹547,000.00INR / Each
Idex Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/PEEK, 1/16" OD Tubing; 1/EA 0201561 Mfr # 7725I-188
COMPARE
Manual Injection Valve, Analytical, Dual Mode with Switch, SS/Ceramic/PEEK, 1/16" OD Tubing; 1/EA 0.0625 SS/Ceramic/PEEK
₹500,000.00INR / Each
Idex Manual Injection Valve, Micro, Dual Mode with Switch, SS, 1/16" OD Tubing; 1/EA 0201562 Mfr # 8125/20UL
COMPARE
Manual Injection Valve, Micro, Dual Mode with Switch, SS, 1/16" OD Tubing; 1/EA 0.0625 SS
₹553,000.00INR / Each
Showing 1- 12 of 12