Idex Manual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA
Representative image only.
Idex Manual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA
Idex | Mfr: 8125| Item: 0201525
Introduce a sample of a solution into a flowing stream of liquid
₹574,000.00INR / Each
₹574,000.00INR / Each
Buy It Together
Current Item
Idex Manual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA
Idex Manual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA
0201525
₹574,000.00INR / Each
+
Idex Rotor Seal for Valve Model 8125, Vespel®; 1/EA
0201564
₹47,800.00INR / Each
=
Total Price 621800.00 ADD 2 ITEMS TO CART
Product Description
Includes
5 µL sample loop
Key Features
  • 2-position, 6-port configuration
  • Front loading with switch
  • SS/ceramic/Vespel® wetted materials
  • 5 µL sample loading volume
  • 10-32 coned ports for 1/50" or 1/16" OD tubing
  • 1 year warranty
More About this Item
Manual injection valves are ideal for applications that require operator control or injection of smaller sample volumes. Injection valves are a type of switching valve usually featuring an attached sample loop (a length of tubing and fittings that match the angle of the valve ports).
Select from single- or dual-mode valves. Single-mode injection valves are configured for rear loading and requires complete filling of the sample loop. Dual-mode injection valves are configured for front loading and allows either partial or complete filling of the loop. Dual-mode valve introduces sample by syringe through the needle port built into the valve shaft. Valves with MBB design creates a continuous flow between the load and inject positions to eliminate pressure transient shock to the system.
1 Year Warranty
Specifications
  • Outlet Port10-32 coned
  • Inlet Port10-32 coned
  • Max Pressure (PSI)5000
  • Max Pressure (bar)345
  • Wetted MaterialsSS/Ceramic/Vespel
  • Tubing OD (in)0.0625
  • DescriptionManual Injection Valve, Micro, Dual Mode with Switch, SS/Ceramic/Vespel®, 1/50" or 1/16" OD Tubing; 1/EA