Idex RheBuild® Kit for Valve Models 8125/8126; 1/EA
Representative image only.
Idex RheBuild® Kit for Valve Models 8125/8126; 1/EA
Idex | Mfr: 8125-999| Item: 0201571
Regular maintenance ensures optimal valve performance
₹77,700.00INR / Each
₹77,700.00INR / Each
Product Description
Key Features
  • Optimize performance of your manual chromatography valves
  • Use with valve models 8125/8126
  • 1 year warranty
More About this Item
RheBulid® kits contains normal wear parts, tools for disassembly/reassembly, and instructions to maintain your valves.
1 Year Warranty
Specifications
  • Compatible WithValve models 8125/8126
  • DescriptionRheBuild® Kit for Valve Models 8125/8126; 1/EA