Idex Rotor Seal for Valve Model 8125, Vespel®; 1/EA
Representative image only.
Idex Rotor Seal for Valve Model 8125, Vespel®; 1/EA
Idex | Mfr: 8125-038| Item: 0201564
Replace rotor seals for longer valve life
₹47,800.00INR / Each
₹47,800.00INR / Each
Product Description
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve model 8125
  • Vespel® seal
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.
1 Year Warranty
Specifications
  • MaterialVespel®
  • Compatible WithValve model 8125
  • DescriptionRotor Seal for Valve Model 8125, Vespel®; 1/EA