Idex Sample Loop Kit for 7725/7725i Valves, SS, 1/16" OD; 1/EA
Representative image only.
Idex Sample Loop Kit for 7725/7725i Valves, SS, 1/16" OD; 1/EA
Idex | Mfr: 7755-060| Item: 0201659
Square-cut, burr-free ends provide a firm, flush connection to valve ports
₹24,800.00INR / Each
₹24,800.00INR / Each
Product Description
Includes
5 µL, 50 µL, and 200 µL volume loops
Key Features
  • 316 stainless steel construction
  • High pressure resistance, low permeability
  • 30° port angle
  • Includes 5, 50, and 200 µL volume loops
  • 1 year warranty
More About this Item
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.
Compatible with injection valve models 7725, 7725i, PR/EV700-100, PR/EV703-100, and MX Module
1 Year Warranty
Specifications
  • Column Inside Diameter (ID) (mm)0.76
  • Wetted Materials316 stainless steel
  • Tubing Outside Diameter (OD) (in)0.0625
  • Total Volume Capacity (µL)200
  • DescriptionSample Loop Kit for 7725/7725i Valves, SS, 1/16" OD; 1/EA