Idex RheBuild® Kit for Valve Models 9725/9725i; 1/EA
Representative image only.
Idex RheBuild® Kit for Valve Models 9725/9725i; 1/EA
Idex | Mfr: 9725-999| Item: 0201325
Regular maintenance ensures optimal valve performance
₹66,200.00INR / Each
₹66,200.00INR / Each
Product Description
Key Features
  • Optimize performance of your manual chromatography valves
  • Use with valve models 9725i/9725i
  • 1 month warranty
More About this Item
RheBulid® kits contains normal wear parts, tools for disassembly/reassembly, and instructions to maintain your valves.
1 Month Warranty
Specifications
  • Compatible WithValve models 9725i/9725i
  • DescriptionRheBuild® Kit for Valve Models 9725/9725i; 1/EA