Idex Rotor Seal for Valve Models 7125 and 7725, Vespel®; 1/EA
Representative image only.
Idex Rotor Seal for Valve Models 7125 and 7725, Vespel®; 1/EA
Idex | Mfr: 7125-047| Item: 0201307
Replace rotor seals for longer valve life
₹39,500.00INR / Each
₹39,500.00INR / Each
Product Description
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7125 and 7725
  • Vespel® seal
  • 1 month warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.
1 Month Warranty
Specifications
  • MaterialVespel®
  • Compatible WithValve models 7125 and 7725
  • DescriptionRotor Seal for Valve Models 7125 and 7725, Vespel®; 1/EA