Idex Stator for Valve Models 7725/7725i, SS; 1/EA
Representative image only.
Idex Stator for Valve Models 7725/7725i, SS; 1/EA
Idex | Mfr: 7725-010| Item: 0201312
Prolong the life of your valves
₹171,000.00INR / Each
₹171,000.00INR / Each
Product Description
Key Features
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve models 7725/7725i
  • 1 month warranty
More About this Item
Optimize performance of your manual chromatography valves with these replacement stators.
1 Month Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithValve models 7725/7725i
  • DescriptionStator for Valve Models 7725/7725i, SS; 1/EA