Environmental Express UltiTube™, Digestion Tubes with Natural Linerless Caps, 50 mL; 150/Box

Representative image only.

Environmental Express UltiTube™, Digestion Tubes with Natural Linerless Caps, 50 mL; 150/Box

Environmental Express | Item: 5320417

Disposable digestion tubes eliminate transfer steps

₹13,200.00INR / Box of 150

₹13,200.00INR / Box of 150

Buy It Together

Current Item

Environmental Express UltiTube™, Digestion Tubes with Natural Linerless Caps, 50 mL; 150/Box
Environmental Express UltiTube™, Digestion Tubes with Natural Linerless Caps, 50 mL; 150/Box
5320417

₹13,200.00INR / Box of 150
+ + + +
Environmental Express Disposable Watch Glass, 50 mL; 1000/Pk
5320104

₹39,000.00INR / Pkg of 1000
+ + + =
Total Price 2336300.00 ADD 8 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Linerless caps, and Certificate of Volume Accuracy and Metals Background Analysis Report for 68 elements to both ppb and ppt (parts per trillion) levels!

Key Features

  • Tubes that act as a digestion vessel or storage container and fit perfectly into the HotBlock and AutoBlock systems
  • Low metals background and EPA approved for digestions
  • Accessories include watch glasses, reflux caps, and transfer/storage racks
  • Redesigned for optimum performance! New certificate of analysis ensures low-levels of 68 elements to ppb and ppt (parts per trillion!) levels—from the industry's cleanest resin Wobble free—deeper bottom dimple provides added stability
  • Model number UCC000-50

More About this Item

Approved for hot plate digestions and certified for low metals background.

Specifications
  • Volume (mL)50
  • Tube MaterialPolypropylene, clean
  • Tube Bottom ShapeFlat
  • Tube ColorTransparent
  • Cap TypeScrew-Thread
  • Cap MaterialUnlined polypropylene
  • Cap colorNatural
  • DescriptionUlti Tube, Digestion Tubes with Natural Linerless Caps, 50 mL; 150/Box
Reviews
Q&A