Environmental Express Disposable Watch Glass, 50 mL; 1000/Pk
Representative image only.
Environmental Express Disposable Watch Glass, 50 mL; 1000/Pk
Environmental Express | Item: 5320104
Optimize performance of your Environmental Express HotBlock digestion tubes
₹39,000.00INR / Pkg of 1000
₹39,000.00INR / Pkg of 1000
Buy It Together
Current Item
Environmental Express Disposable Watch Glass, 50 mL; 1000/Pk
Environmental Express Disposable Watch Glass, 50 mL; 1000/Pk
5320104
₹39,000.00INR / Pkg of 1000
+ + + + =
Total Price 2027900.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Watch glasses to cover your tubes during digestion
  • Transfer racks to avoid spillage
  • Caps to close tubes
  • 1 year warranty
  • Model number SC505
More About this Item
Choose from a wide range of accessories and replacement parts for your digestion tubes.
Watch glasses are made from metals-free polypropylene and are ideal for digestions where reagents need to be added throughout the heating process. Watch glasses have a center stem for easy handling and to decrease the chances of contamination.
1 Year Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible With50 mL digestion tubes
  • DescriptionDisposable Watch Glass, 50 mL; 1000/Pk