Environmental Express C6010 TKN Digestion Tube, Threaded; 1/Ea
Representative image only.
Environmental Express C6010 TKN Digestion Tube, Threaded; 1/Ea
Environmental Express | Item: 3520572
Optimize your TKN analysis for better results
₹14,500.00INR / Each
₹14,500.00INR / Each
Buy It Together
Current Item
Environmental Express C6010 TKN Digestion Tube, Threaded; 1/Ea
Environmental Express C6010 TKN Digestion Tube, Threaded; 1/Ea
3520572
₹14,500.00INR / Each
+ + =
Total Price 65600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Interchangeable glassware works with SimpleDist and TKN HotBLock systems
  • Maximum heat transfer with flat-bottom, borosilicate glassware
  • Ideal for reflux and digestion methods
  • Model number C6010
More About this Item
Teardrop stoppers are ideal for the capture and condesation of digestion fumes, preventing cross contamination (sold separately).
Specifications
  • Capacity (mL)50
  • DescriptionThreaded Digestion Tube, EA