Environmental Express TKN6012 Teardrop Stopper for TKN Digestion Tubes; 1/Ea
Representative image only.
Environmental Express TKN6012 Teardrop Stopper for TKN Digestion Tubes; 1/Ea
Environmental Express | Item: 3520574
Capture condensation
₹11,200.00INR / Each
₹11,200.00INR / Each
Buy It Together
Current Item
Environmental Express TKN6012 Teardrop Stopper for TKN Digestion Tubes; 1/Ea
Environmental Express TKN6012 Teardrop Stopper for TKN Digestion Tubes; 1/Ea
3520574
₹11,200.00INR / Each
+ =
Total Price 25700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Prevent cross-contamination
  • Model number TKN6012
More About this Item
Teardrop stoppers are ideal for the capture and condesation of digestion fumes, preventing cross contamination.
Specifications
  • DescriptionTeardrop Stopper for Digestion Tubes, EA