Cole-Parmer Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 216 mL/min. (Air), 3.0 mL/min. (H2O), Cartridge Valve
Representative image only.
Cole-Parmer Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 216 mL/min. (Air), 3.0 mL/min. (H2O), Cartridge Valve
Cole-Parmer | Item: 1530975
Ideal for precise low flow rate measurements
₹26,000.00INR / Each
for an additional
₹49,800.00INR / Each
for an additional
₹63,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹26,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 216 mL/min. (Air), 3.0 mL/min. (H2O), Cartridge Valve
Cole-Parmer Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 216 mL/min. (Air), 3.0 mL/min. (H2O), Cartridge Valve
1530975
₹26,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
1708000
₹49,800.00INR / Each
+ + + + =
Total Price 2946800.00 ADD 6 ITEMS TO CART
Product Description
Includes
Cartridge valve and mounting fittings.
Key Features
  • Corrosion-resistant aluminum fittings ensure long-term reliability
  • Easy installation and exchange of flow tubes
  • Non-rotating adapter prevents tubes from turning
  • Panel mounting with hex nuts makes installation simple and secure
  • Chemical compatibility
  • 1 year warranty
More About this Item
These single flow tube variable area flowmeters with cartridge valves are designed for precise low flow rate measurements of air and water in mL/minute at a given set of pressure and temperature parameters. The Optigrad™ 65 mm universal millimeter scale minimizes parallax and eye fatigue. Particularly suitable for metering carrier gases in chromatography, indicating and controlling gases in manufacturing processes, liquid and gas measurement in laboratories, and pilot plants.
Built-in high-resolution control valves offer high sensitivity control and resolution in conjunction with metering tubes of very low flow rates. Other features include standard mounting fittings, side plates, thick protective magnifying front shield and back plate, and flow tubes.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
1 Year Warranty
Specifications
  • Accuracy±5% of full scale reading
  • Repeatability±0.25%
  • Width (InchShort)1 1/4
  • Number of Channels1
  • Height (InchShort)5 1/2
  • Max Pressure (psig)200
  • Max Air Flow Rate (mLmin)216
  • Max Water Flow Rate (mLmin)3
  • Material Flow TubeBorosilicate glass
  • Material FittingAluminum
  • Material O-RingBuna®
  • Material FloatGlass
  • Material HousingAluminum, black anodized
  • ValveCartridge
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))1/8
  • Fluid TypeWater
  • Gas TypeAir
  • DescriptionVariable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 216 mL/min. (Air), 3.0 mL/min. (H2O), Cartridge Valve