Alicat Whisper™ MCW-Series Low-Pressure Drop Mass Flow Controllers, SCCM
Accurately measures and controls real-time mass and volumetric flow readings with minimal pressure drop
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ideal for systems with limited source pressure
  • Less than one millisecond sensor response time
  • IP40 rating
These low-pressure drop mass flow controllers, featuring a fast control response time of just 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. Controllers create minimal pressure drop, making them ideal for systems with limited source pressure. They are capable of measuring and controlling flow generated by both positive pressure and suction. Real-time monitoring of flow, pressure, and temperature is available directly on the display screen.
The controllers come with 98 pre-loaded gas calibrations, which can be seamlessly switched while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure, with flow rates displayed in standard cubic centimeters per minute (SCCM). Additional key features include an LCD display with backlight, a data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
283,000.00 - ₹379,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Process Connection Accuracy
Availability Pricing
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 0.5 SCCM 6780382 Mfr # MCW-0.5SCCM-D
COMPARE
0.5 M5 female thread (10-32 compatible) ±0.8% of reading + 0.2% of full scale
₹379,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 1 SCCM 6780383 Mfr # MCW-1SCCM-D
COMPARE
1 M5 female thread (10-32 compatible) ±0.8% of reading + 0.2% of full scale
₹379,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 2 SCCM 6780384 Mfr # MCW-2SCCM-D
COMPARE
2 M5 female thread (10-32 compatible) ±0.8% of reading + 0.2% of full scale
₹379,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 5 SCCM 6780385 Mfr # MCW-5SCCM-D
COMPARE
5 M5 female thread (10-32 compatible) ±0.8% of reading + 0.2% of full scale
₹283,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 10 SCCM 6780386 Mfr # MCW-10SCCM-D
COMPARE
10 M5 female thread (10-32 compatible) ±0.75% of reading or ±0.1% of full scale, whichever is greater
₹283,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 20 SCCM 6780387 Mfr # MCW-20SCCM-D
COMPARE
20 M5 female thread (10-32 compatible) ±0.75% of reading or ±0.1% of full scale, whichever is greater
₹283,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 50 SCCM 6780388 Mfr # MCW-50SCCM-D
COMPARE
50 1/8" NPT(F) ±0.75% of reading or ±0.1% of full scale, whichever is greater
₹283,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 100 SCCM 6780389 Mfr # MCW-100SCCM-D
COMPARE
100 ±0.75% of reading or ±0.1% of full scale, whichever is greater
₹283,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 200 SCCM 6780390 Mfr # MCW-200SCCM-D
COMPARE
200 1/8" NPT(F) ±0.75% of reading or ±0.1% of full scale, whichever is greater
₹283,000.00INR / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 500 SCCM 6780391 Mfr # MCW-500SCCM-D
COMPARE
500 1/8" NPT(F) ±0.75% of reading or ±0.1% of full scale, whichever is greater
₹283,000.00INR / Each
Showing 1- 10 of 10