InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
Representative image only.
InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
InnoCal | Item: 1708000
With the variety of flowmeter types—from differential pressure to magnetic to ultrasonic—it helps to have highly trained metrologists calibrating the meters for optimal performance
₹49,800.00INR / Each
₹49,800.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
1708000
₹49,800.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 2 SLPM
1530905
₹229,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 200 SCCM
1530902
₹222,000.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 1 SLPM
1530881
₹169,000.00INR / Each
+ =
Total Price 730200.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s), and test uncertainty ratios (TUR’s) at no additional cost
  • This NIST-traceable calibration allows you to confirm that the specific asset you purchase meets the manufacturer’s published specification
  • EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant
  • Testing includes measurement at 5 to 10 points—depending on the meter–equidistant across the range of the flowmeter
  • Testing range  0-500 LPM air flow
  • Calibration service for an Air/gas flowmeter
  • Testing range 0.1 to 80,000 sccm at uncertainties as low as (K=2) ±0.2% of reading
  • Testing includes measurement at five to seven points — depending on the meter — equidistant across the range of the flowmeter
  • Certificate includes measured data, graphs, and correlations (if required) to meet the most stringent quality and scientific requirements
More About this Item
At InnoCal, our computerized, automated standards allow us to perform NIST-traceable mass and correlated flow measurements using air or nitrogen gas. When correlation to other gases is needed, our metrologists will provide you with correlated data or a gas correlation table.
Typical brands that we calibrate include Cole-Parmer, Alicat, Aalborg Instruments, Gilmont, Sierra Instruments, Bios International, and many more.
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A.
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory.
NIST Traceable Calibration
Specifications
  • Calibration TypeAir/Gas flow
  • Calibration Test Points5 to 10 points—depending on the meter
  • Calibration Test Range0-500 LPM air flow
  • DescriptionNIST-Traceable Calibration; Air/Gas Flowmeter