Alicat M-Series Mass Flowmeters with Digital Display, SLPM
Provides fast, stable, repeatable flow rate control in milliseconds
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.6% of reading
  • A2LA 17025 Accredited calibration is available as an additional service and cost through our InnoCal Metrology Lab
  • Lifetime warranty
These mass flowmeters, with a fast control response time of 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They come with 98 pre-loaded gas calibrations, which can be switched seamlessly while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure with flow rates displayed in standard liters per minute (SLPM). Key features include a backlit monochrome display, data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Accuracy Repeatability
Availability Pricing
Alicat Mass Flowmeter with Display, 0 - 1 SLPM 1530881 Mfr # M-1SLPM-D
COMPARE
1 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
₹169,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 2 SLPM 1530882 Mfr # M-2SLPM-D
COMPARE
2 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
₹169,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 5 SLPM 1530883 Mfr # M-5SLPM-D
COMPARE
5 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
₹169,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 10 SLPM 1530884 Mfr # M-10SLPM-D
COMPARE
10 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
₹169,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 20 SLPM 1530885 Mfr # M-20SLPM-D
COMPARE
20 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
₹169,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 50 SLPM 1530886 Mfr # M-50SLPM-D
COMPARE
50 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹187,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 100 SLPM 1530887 Mfr # M-100SLPM-D
COMPARE
100 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹187,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 250 SLPM 1530888 Mfr # M-250SLPM-D
COMPARE
250 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹265,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 500 SLPM 1530889 Mfr # M-500SLPM-D
COMPARE
500 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹256,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 1000 SLPM 1530890 Mfr # M-1000SLPM-D
COMPARE
1000 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹340,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 2000 SLPM 1530891 Mfr # M-2000SLPM-D
COMPARE
2000 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹470,000.00INR / Each
Alicat Mass Flowmeter with Display, 0 - 3000 SLPM 1530892 Mfr # M-3000SLPM-D
COMPARE
3000 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹537,000.00INR / Each
Alicat Mass Flowmeter with Display, 0-5000 SLPM 1530893 Mfr # M-5000SLPM-D
COMPARE
5000 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
₹745,000.00INR / Each
Showing 1- 13 of 13