Ohaus Frontier High-Volume Refrigerated Centrifuges
Suitable for routine and research applications in clinical, biotech, and industrial labs
  • Automatic rotor identification
  • Simple one-hand operation
  • Intuitive, easy-to-read backlit LCD display
  • Store up to 99 runs for easy recall
  • Motorized lid-locking system prevents lid from opening during operation
  • Imbalance sensor stops rotor when imbalance is detected
  • Integrated wheels make it easy to move around
These high-speed, high-capacity refrigerated centrifuges offer automatic rotor recognition, robust safety features, and versatile rotor compatibility for maximum lab flexibility. With a capacity of up to 4 × 1000 mL, they can spin a wide range of bottles, tubes, and plates using fixed-angle or swing-out rotors. A powerful motor delivers precise separations, reaching speeds up to 16,010 rpm and forces up to 26,361 × g. The intuitive design allows easy loading and unloading, quick access to settings, and real-time parameter adjustments without stopping the run.
Key features include one-hand operation, backlit LCD display, and easy-to-use rotary knob for quick adjustments. Select from 10 acceleration and 10 deceleration rates to suit your application. Store up to 99 runs with relevant parameters, including the used rotors, to easily repeat the procedures and save time. A self-diagnostic program records usage and error history for easy maintenance and reduced downtime. Constructed of chemical and corrosion-resistant stainless steel ensures durability in lab use.
Safety highlights include over-speed protection, a motorized lid-locking system that prevents the lid from opening during operation, and an imbalance sensor that stops the rotor when imbalanced load is detected.
2,262,000.00 - ₹2,458,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Max Speed (rpm) Refrigerated
Availability Pricing
Ohaus Frontier FC5917RF High-Volume Refrigerated Centrifuge, 16,010 rpm, 230V 8524152 Mfr # 83042055
COMPARE
1000 16010 Yes
₹2,458,000.00INR / Each
Ohaus Frontier FC5917RF High-Volume Short Refrigerated Centrifuge, 16,010 rpm, 230V 8524153 Mfr # 83042056
COMPARE
1000 16010 Yes
₹2,262,000.00INR / Each