CPC (Colder) Tentube™ Quick-Disconnect Fitting, Hose Barb Female Body, Acetal, Non-Valved, 1/8" ID
Representative image only.
CPC (Colder) Tentube™ Quick-Disconnect Fitting, Hose Barb Female Body, Acetal, Non-Valved, 1/8" ID
CPC (Colder) | Mfr: TFB1002| Item: 5013682
Quickly connect up to 10 fluid lines
₹3,100.00INR / Each
₹3,100.00INR / Each
Buy It Together
Current Item
CPC (Colder) Tentube™ Quick-Disconnect Fitting, Hose Barb Female Body, Acetal, Non-Valved, 1/8" ID
CPC (Colder) Tentube™ Quick-Disconnect Fitting, Hose Barb Female Body, Acetal, Non-Valved, 1/8" ID
5013682
₹3,100.00INR / Each
+ + + + =
Total Price 2904300.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Multi-line connections for fast, efficient operation
  • Separate flow paths all in one fitting
  • Insert is keyed to body to ensure correct line orientation
  • Ideal for tubing sizes from 1/16” to 1/8” ID
More About this Item
The CPC (Colder) Tentube quick-disconnect hose barb body allows you to easily connect and disconnect up to 10 tube lines with the use of a simple slide latch. It features in-line configurations for maximum design flexibility.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialNylon
  • Valve MaterialAcetal
  • Valve Spring MaterialStainless Steel
  • O-Ring MaterialBuna-N
  • Connection TypeQuick-Disconnect Hose Barb Body, Non-Valved
  • Number Of Ports10
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Flow PatternStraight
  • ColorBlack
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)100
  • DescriptionQuick-Disconnect Fitting, Hose Barb Female Body, Acetal, Non-Valved, 1/8" ID