Cole-Parmer Nylon Membrane Filters; 0.45µm Pore, 25mm Dia, 100/Pk

Representative image only.

Cole-Parmer Nylon Membrane Filters; 0.45µm Pore, 25mm Dia, 100/Pk

Cole-Parmer | Item: 1594525

Filter aqueous and organic solvents without prewetting

₹1,200.00INR / Each

₹1,200.00INR / Each

Buy It Together

Current Item

Cole-Parmer Nylon Membrane Filters; 0.45µm Pore, 25mm Dia, 100/Pk
Cole-Parmer Nylon Membrane Filters; 0.45µm Pore, 25mm Dia, 100/Pk
1594525

₹1,200.00INR / Each
+
Cole-Parmer Nylon Syringe Filters, 0.45 um, 25 mm Dia; 100/Box
0291514

₹36,800.00INR / Box of 100
+
Cole-Parmer Nylon (Polyamide) Membranes, 0.2 um, 13 mm Dia; 100/Pk
0291630

₹41,800.00INR / Pkg of 100
+ + + + +
Cole-Parmer PTFE Membrane, 47 mm, 0.2 µm, 100/pk
3622924

₹71,200.00INR / Pkg of 100
+
Cole-Parmer Nylon Membrane Filters, 0.1 µm, 25 mm; 100/Pk
3623048

₹82,000.00INR / Pkg of 100
+ =
Total Price 303100.00 ADD 10 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Use for HPLC sample preparation, to sterilize biological solutions/buffers, and vacuum degassing

More About this Item

Nonsterile nylon membranes are inherently hydrophilic and are compatible with many aqueous and organic solvents and solutions making them suitable for HPLC sample preparation, sterilizing biological solutions or buffers, and vacuum degassing. Membranes may be exposed to temperatures up to 75°C (167°F), and can withstand up to five autoclave cycles at 121°C for 30 minutes. Additional variations are available; call us for details.

ISO9001 Certified Supplier
Specifications
  • MaterialNylon
  • Pore Size (µm)0.45
  • Diameter (mm)25
  • DescriptionNylon Membrane Filters; 0.45µm Pore, 25mm Dia, 100/Pk
Reviews
Q&A