Advantec Sterile Mixed Cellulose Ester (MCE) Filter Membranes

Presterilized membranes in convenient single-, multi-, or continuous-feed packs
  • Unique high porosity provides superior flow rate
  • Can be safely exposed to temperatures up to 130°C (266°F)
Mixed cellulose ester membranes are suitable for microbiological analysis of water, wastewater, pharmaceuticals, or beverages. Offered with or without grid lines; grid lines facilitate counting colonies on the membrane surface. Grid squares measure 3.1 mm and represent 1/100 of the area of a 47-mm membrane. Black membranes maximize contrast between colonies and the filter; green membrane enables viewing of black, white, and colorless colonies on a single filter. Presterilized by EtO, these 47-mm diameter filters are available in envelopes of 10, individually packaged, or now individually wrapped continuous feed. Order membranes with absorbent pad for use with broth media.
Call For Price

16 variations of this product are available.

See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
Advantec A045D047A Sterile MCE Gridded Filters w/Pads, 0.45um, 47mm Dia; 10x10/Pk 0664408 Mfr # A045D047A
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A045D047Y Sterile MCE Gridded Filters, 0.45um, 47mm Dia; 20x10/Pk 0664410 Mfr # A045D047Y
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A045H047A Sterile MCE Gridded Filters, 0.45um, 47mm Dia; 100/Pk 0664414 Mfr # A045H047A
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A020F047A Sterile MCE Gridded Filters w/Pads, 0.20um, 47mm Dia; 100/Pk 0664419 Mfr # A020F047A
COMPARE
47 Mixed Cellulose Ester 0.2
Call For Price
Advantec A020G047A Sterile MCE Filters, 0.20um, 47mm Dia; 100/Pk 0664421 Mfr # A020G047A
COMPARE
47 Mixed Cellulose Ester 0.2
Call For Price
Advantec A045G047A Sterile MCE Filters, 0.45um, 47mm Dia; 100/Pk 0664427 Mfr # A045G047A
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A045H047W Sterile MCE Gridded Filters, 0.45um, 47mm Dia; 1000/Pk 0664431 Mfr # A045H047W
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A045H047Y Sterile MCE Gridded Filters, 0.45um, 47mm Dia; 200/Pk 0664433 Mfr # A045H047Y
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A045R047A Sterile MCE Gridded Black Filters w/o Pads, 0.45um, 47mm; 100/Pk 0664439 Mfr # A045R047A
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A080F047A Sterile MCE Gridded Filters w/Pads, 0.80um, 47mm Dia; 100/Pk 0664445 Mfr # A080F047A
COMPARE
47 Mixed Cellulose Ester 0.8
Call For Price
Advantec A080G047A Sterile MCE Filters, 0.80um, 47mm Dia; 100/Pk 0664447 Mfr # A080G047A
COMPARE
47 Mixed Cellulose Ester 0.8
Call For Price
Advantec A080H047A Sterile MCE Gridded Filters, 0.80um, 47mm Dia; 100/Pk 0664449 Mfr # A080H047A
COMPARE
47 Mixed Cellulose Ester 0.8
Call For Price
Advantec A080H047W Sterile MCE Gridded Filters, 0.80um, 47mm Dia; 1000/Pk 0664452 Mfr # A080H047W
COMPARE
47 Mixed Cellulose Ester 0.8
Call For Price
Advantec A045W047A Sterile MCE Gridded Green Filters, 0.45um, 47mm Dia; 100/Pk 0664454 Mfr # A045W047A
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Advantec A100H047A Sterile Mixed Cellulose Ester Filter Membranes 1.0 WG IND 47MM 100/PK 0664455 Mfr # A100H047A
COMPARE
47 Mixed Cellulose Ester 0.8
Call For Price
Advantec Sterile MCE Filters, 0.45um, 47mm; 150/Pk Continuous Feed 1664414 Mfr # A045H047Z-P
COMPARE
47 Mixed Cellulose Ester 0.45
Call For Price
Showing 1- 16 of 16

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat