Advantec A020G047A Sterile MCE Filters, 0.20um, 47mm Dia; 100/Pk

Representative image only.

Advantec A020G047A Sterile MCE Filters, 0.20um, 47mm Dia; 100/Pk

Advantec | Mfr: A020G047A| Item: 0664421

Presterilized membranes in convenient single-, multi-, or continuous-feed packs

Call For Price

Call For Price

Buy It Together

Current Item

Advantec A020G047A Sterile MCE Filters, 0.20um, 47mm Dia; 100/Pk
Advantec A020G047A Sterile MCE Filters, 0.20um, 47mm Dia; 100/Pk
0664421

Call For Price
+ + + =
Total Price 4200.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Unique high porosity provides superior flow rate
  • Can be safely exposed to temperatures up to 130°C (266°F)
  • 1 year warranty

More About this Item

Mixed cellulose ester membranes are suitable for microbiological analysis of water, wastewater, pharmaceuticals, or beverages. Offered with or without grid lines; grid lines facilitate counting colonies on the membrane surface. Grid squares measure 3.1 mm and represent 1/100 of the area of a 47-mm membrane. Black membranes maximize contrast between colonies and the filter; green membrane enables viewing of black, white, and colorless colonies on a single filter. Presterilized by EtO, these 47-mm diameter filters are available in envelopes of 10, individually packaged, or now individually wrapped continuous feed. Order membranes with absorbent pad for use with broth media.

1 Year Warranty
Specifications
  • MaterialMixed Cellulose Ester
  • Pore Size (µm)0.2
  • Diameter (mm)47
  • SurfacePlain
  • Absorbent PadNot included
  • DescriptionSterile MCE Filters, 0.20um, 47mm Dia; 100/Pk
  • Warranty1 year
Reviews
Q&A