Cole-Parmer® Nylon Syringe Filters
Hydrophilic nylon membranes have extremely low levels of extractables and contain no wetting agents
  • They offer good chemical resistance and are ideal for aqueous and most organic samples
Filters are Triton-free and have a bidirectional membrane support and luer lock inlet/luer slip outlet for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. They are certified nonpyrogenic and noncytotoxic.
14,200.00 - ₹26,000.00INR / Box of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
25 0.45 Yes
₹15,900.00INR / Box of 50
COMPARE
25 0.2 Yes
₹17,100.00INR / Box of 50
COMPARE
25 0.45 No
₹26,000.00INR / Box of 100
COMPARE
25 0.02 No
₹14,200.00INR / Box of 100