Cole-Parmer® Nylon (Polyamide) Membranes
Ideal for filtering or sterilizing nonprotein-containing biological solutions
  • High protein binding
  • Withstand repeated steam sterilization
Inherently hydrophilic membranes do not require prewetting — use to filter aqueous solutions and organic solvents.
7,200.00 - ₹86,000.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
13 0.2
₹41,800.00INR / Pkg of 100
COMPARE
47 0.2
₹71,000.00INR / Pkg of 100
COMPARE
90 0.2
₹48,600.00INR / Pkg of 25
COMPARE
142 0.2
₹86,000.00INR / Pkg of 25
COMPARE
13 0.45
₹38,000.00INR / Pkg of 100
COMPARE
47 0.45
₹64,600.00INR / Pkg of 100
COMPARE
90 0.45
₹36,600.00INR / Pkg of 25
COMPARE
13 0.2
₹7,200.00INR / Pkg of 100
COMPARE
25 0.2
₹31,300.00INR / Pkg of 100
COMPARE
47 0.2
₹25,300.00INR / Pkg of 100
COMPARE
90 0.2
₹28,400.00INR / Pkg of 25
COMPARE
25 0.45
₹31,300.00INR / Pkg of 100
COMPARE
47 0.45
₹34,400.00INR / Pkg of 100
COMPARE
90 0.45
₹20,700.00INR / Pkg of 25
Showing 1- 14 of 14