Cole-Parmer® PTFE Membranes
Unaffected by even the most corrosive compounds, including concentrated acids, alkaline, and propellants
  • Durable laminated PTFE membranes
  • Prewet with acetone or methanol for aqueous solutions
Membranes withstand temperatures from –100 to 130°C (–148 to 266°F). Use these hydrophobic membranes for sterile venting and filtration of gas, air steams, and organic solvents.
14,400.00 - ₹61,200.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
13 Hydrophobic PTFE 0.2
₹14,900.00INR / Pkg of 100
COMPARE
25 Hydrophobic PTFE 0.2
₹32,000.00INR / Pkg of 100
COMPARE
47 Hydrophobic PTFE 0.2
₹49,100.00INR / Pkg of 100
COMPARE
142 Hydrophobic PTFE 0.2
₹61,200.00INR / Pkg of 25
COMPARE
13 Hydrophobic PTFE 0.45
₹14,400.00INR / Pkg of 100
COMPARE
25 Hydrophobic PTFE 0.45
₹31,500.00INR / Pkg of 100
COMPARE
47 Hydrophobic PTFE 0.45
₹48,000.00INR / Pkg of 100
COMPARE
90 Hydrophobic PTFE 0.45
₹35,900.00INR / Pkg of 25
COMPARE
142 Hydrophobic PTFE 0.45
₹59,400.00INR / Pkg of 25
COMPARE
13 Hydrophobic PTFE 1.2
₹14,400.00INR / Pkg of 100
COMPARE
25 Hydrophobic PTFE 1.2
₹37,500.00INR / Pkg of 100
COMPARE
47 Hydrophobic PTFE 1.2
₹49,500.00INR / Pkg of 100
COMPARE
90 Hydrophobic PTFE 1.2
₹40,300.00INR / Pkg of 25
COMPARE
142 Hydrophobic PTFE 1.2
₹61,200.00INR / Pkg of 25
Showing 1- 14 of 14