Chemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea

Representative image only.

Chemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea

| Item: 0671580

Resistant to mild acids, alkaline solutions and many reagents.

₹7,400.00INR / Each

₹7,400.00INR / Each

Buy It Together

Current Item

Chemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea
Chemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea
0671580

₹7,400.00INR / Each
+ + + +
Cole-Parmer Stainless steel utility tray, 10"L x 6-1/2"W
0727711

₹9,300.00INR / Each
+ +
Cole-Parmer Airflow Meter Vaneometer
7890012

₹22,900.00INR / Each
+ + =
Total Price 87900.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Withstand temperatures from –40 to 250°F (–40 to 121°C)
  • Contain spills with straight edge pans

More About this Item

Constructed of Thermoset polyester resin, these white trays are ideal for carrying, storing, and cleaning of lab product. Trays withstands temperatures from 40°C to 121° (-40°F to 250°F). Pans with straight edges are ideal for spill containment.

Specifications
  • ColorWhite
  • MaterialPolyester
  • Overall Length (in)13 1/4
  • Overall Length (cm)33.7
  • Overall Width (in)10 5/8
  • Overall Width (cm)27
  • Overall Height (in)1
  • Overall Height (cm)2.5
  • DescriptionChemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea
Reviews
Q&A