Chemical Resistant Trays
Resistant to mild acids, alkaline solutions and many reagents.
  • Withstand temperatures from –40 to 250°F (–40 to 121°C)
  • Contain spills with straight edge pans
Constructed of Thermoset polyester resin, these white trays are ideal for carrying, storing, and cleaning of lab product. Trays withstands temperatures from 40°C to 121° (-40°F to 250°F). Pans with straight edges are ideal for spill containment.
6,200.00 - ₹12,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (in) Overall Length (in)
Availability Pricing
COMPARE
1 13 1/4
₹6,200.00INR / Each
COMPARE
1 18
₹7,300.00INR / Each
COMPARE
1 20 3/8
₹8,700.00INR / Each
COMPARE
1 1/8 25 1/4
₹12,800.00INR / Each
COMPARE
1 1/4 19 1/2
₹8,700.00INR / Each
COMPARE
2 3/4 31
₹12,900.00INR / Each