Alicat Mass Flowmeter with Display, 0 - 5 SCCM, Battery Operated
Representative image only.
Alicat Mass Flowmeter with Display, 0 - 5 SCCM, Battery Operated
Alicat | Mfr: MB-5SCCM-D| Item: 6780272
Provides accurate, real-time mass and volumetric flow readings
₹198,000.00INR / Each
for an additional
₹63,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹198,000.00INR / Each
Buy It Together
Current Item
Alicat Mass Flowmeter with Display, 0 - 5 SCCM, Battery Operated
Alicat Mass Flowmeter with Display, 0 - 5 SCCM, Battery Operated
6780272
₹198,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 200 SCCM
1530902
₹222,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 20 SLPM
1530908
₹222,000.00INR / Each
+
Alicat Liquid Flowmeter with Display, 0 - 0.5 CCM
1530917
₹249,000.00INR / Each
+
Alicat Liquid Flowmeter with Display, 0 - 1 LPM
1530927
₹181,000.00INR / Each
+
Alicat Single Ended Mini-Din Cable, 8-Pin Conductor, 6'
1530938
₹4,000.00INR / Each
+ + + =
Total Price 1728000.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Built-in rechargeable battery and micro-USB to USB with wall adapter for power and communication.
Key Features
  • Built-in rechargeable battery with up to 18 hours life
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • IP40 rating
  • Limited lifetime warranty
More About this Item
These portable mass flowmeters, with a fast control response time of 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They come with 98 pre-loaded gas calibrations, which can be switched seamlessly while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure with flow rates displayed in standard cubic centimeters per minute (SCCM). Key features include a rechargeable battery, backlit monochrome display, data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Conformite EuropeenneCanadian Standards AssociationLifetime Warranty
Specifications
  • Accuracy±0.6% of reading
  • Repeatability±0.2% of reading + 0.02% of full scale
  • Width (InchShort)2 3/8
  • Height (InchShort)5 35/64
  • Max Pressure (PSI)75
  • Min Flow Rate (SCCM)0
  • Max Flow Rate (SCCM)5
  • Wetted MaterialsStainlesss steel, fluoroelastomers, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon
  • Min Operating Temperature (° F)14
  • Min Operating Temperature (° C)-10
  • Max Operating Temperature (° F)140
  • Max Operating Temperature (° C)60
  • Process Connection (NPT(F))1/8
  • Process ConnectionM5 female (10-32 compatible), shipped with Buna-N O-ring face seal to 1/8″ NPT female fittings
  • Response Time (msec)10
  • Power VDC5
  • BatteryRechargeable battery
  • Battery LifeUp to 18 hours
  • Electrical Connections6-pin locking, 8-pin mini-DIN, 8-pin M12, 9-pin DB-9, 15-pin DB-15
  • Display TypeBack-lit monochrome
  • CommunicationsRS-232
  • DescriptionMass Flowmeter with Display, 0 - 5 SCCM, Battery
  • WarrantyLifetime