Alicat LB-Series Liquid Flowmeters with Digital Display, Battery Operated, LPM
Accurately measures flow rate, pressure, and temperature of liquids in real-time
  • Built-in rechargeable battery with up to18 hours life
  • No warm-up required
  • Thirty millisecond measurement response time
  • Steady state control 2-100% of full scale
  • NIST-traceable accuracy up to ±2% of full scale
  • IP40 rating
  • Lifetime warranty
These portable, battery-operated liquid flowmeters are designed for high-performance applications where precise monitoring of fluid parameters is crucial. With a rapid control response time of just 30 milliseconds (ms), they provide real-time measurements of several key fluid properties, including flow rate, pressure, and temperature. This allows for accurate and stable control across a variety of liquids, such as water, methanol, ethanol, ammonia, dimethyl ether, iso-pentane, thiophene, iso-octane, and heptane.
The flowmeters offer an intuitive monochrome display where all the measurement data—flow rate, pressure, and temperature—can be easily viewed at the same time. The measurements are presented in liters per minute (LPM), ensuring clarity for users working with precise fluid quantities.
Additionally, the flowmeters come with convenient bleed screws, enabling users to remove air pockets from the system quickly, ensuring optimal performance. For enhanced data management, the meters also feature a data I/O port, which allows for seamless logging of data to your computer, making it easy to monitor and analyze flow performance over time. These features make the flowmeter a reliable solution for various industrial applications requiring consistent and accurate fluid measurement.
210,000.00 - ₹278,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Accuracy Repeatability
Availability Pricing
Alicat Liquid Flowmeter with Display, 0 - 1 LPM, Battery Operated 6780440 Mfr # LB-1LPM-D
COMPARE
1 ±2% of full scale ±0.2% of full scale
₹210,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 2 LPM, Battery Operated 6780441 Mfr # LB-2LPM-D
COMPARE
2 ±2% of full scale ±0.2% of full scale
₹210,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 5 LPM, Battery Operated 6780442 Mfr # LB-5LPM-D
COMPARE
5 ±2% of full scale ±0.2% of full scale
₹248,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 10 LPM, Battery Operated 6780443 Mfr # LB-10LPM-D
COMPARE
10 ±2% of full scale ±0.2% of full scale
₹278,000.00INR / Each