Alicat L-Series Liquid Flowmeters with Digital Display, CCM
Accurately measures flow rate, pressure, and temperature of liquids in real-time
  • No warm-up required
  • Thirty millisecond measurement response time
  • Steady state control 2-100% of full scale
  • NIST-traceable accuracy up to ±2% of full scale
  • IP40 rating
  • Lifetime warranty
These liquid flowmeters are designed for high-performance applications where precise monitoring of fluid parameters is crucial. With a rapid control response time of just 30 milliseconds (ms), they provide real-time measurements of several key fluid properties, including flow rate, pressure, and temperature. This allows for accurate and stable control across a variety of liquids, such as water, methanol, ethanol, ammonia, dimethyl ether, iso-pentane, thiophene, iso-octane, and heptane.
The flowmeters offer an intuitive monochrome display where all the measurement data—flow rate, pressure, and temperature—can be easily viewed at the same time. The measurements are presented in cubic centimeters per minute (CCM), ensuring clarity for users working with precise fluid quantities.
Additionally, the flowmeters come with convenient bleed screws, enabling users to remove air pockets from the system quickly, ensuring optimal performance. For enhanced data management, the meters also feature a data I/O port, which allows for seamless logging of data to your computer, making it easy to monitor and analyze flow performance over time. These features make the flowmeter a reliable solution for various industrial applications requiring consistent and accurate fluid measurement.
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Accuracy Repeatability
Availability Pricing
Alicat Liquid Flowmeter with Display, 0 - 0.5 CCM 1530917 Mfr # L-0.5CCM-D
COMPARE
0.5 ±2% of full scale ±0.2% of full scale
₹249,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 1 CCM 1530918 Mfr # L-1CCM-D
COMPARE
1 ±2% of full scale ±0.2% of full scale
₹249,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 2 CCM 1530919 Mfr # L-2CCM-D
COMPARE
2 ±2% of full scale ±0.2% of full scale
₹249,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 5 CCM 1530920 Mfr # L-5CCM-D
COMPARE
5 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 10 CCM 1530921 Mfr # L-10CCM-D
COMPARE
10 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 20 CCM 1530922 Mfr # L-20CCM-D
COMPARE
20 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 50 CCM 1530923 Mfr # L-50CCM-D
COMPARE
50 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 100 CCM 1530924 Mfr # L-100CCM-D
COMPARE
100 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 200 CCM 1530925 Mfr # L-200CCM-D
COMPARE
200 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Alicat Liquid Flowmeter with Display, 0 - 500 CCM 1530926 Mfr # L-500CCM-D
COMPARE
500 ±2% of full scale ±0.2% of full scale
₹181,000.00INR / Each
Showing 1- 10 of 10