Alicat Single Ended Mini-Din Cable, 8-Pin Conductor, 6'
Representative image only.
Alicat Single Ended Mini-Din Cable, 8-Pin Conductor, 6'
Alicat | Mfr: DC-61| Item: 1530938
Keep your mass flowmeters up and running
₹4,000.00INR / Each
₹4,000.00INR / Each
Buy It Together
Current Item
Alicat Single Ended Mini-Din Cable, 8-Pin Conductor, 6'
Alicat Single Ended Mini-Din Cable, 8-Pin Conductor, 6'
1530938
₹4,000.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 500 SCCM
1530880
₹169,000.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 1 SLPM
1530881
₹169,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 200 SCCM
1530902
₹222,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 20 SLPM
1530908
₹222,000.00INR / Each
+ +
Alicat Liquid Flow Controller with Display, 0 - 5 CCM
1530931
₹258,000.00INR / Each
+ + + =
Total Price 1903600.00 ADD 10 ITEMS TO CART
Product Description
More About this Item
Keep your Alicat mass flowmeters running properly with the following power cable and adapter.
This single-ended DC series cable has a male 8-pin connector on one side and a blunt cut cable on the other. It is capable of carrying power, analog, and digital signals.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Compatible WithAlicat mass flowmeters and controllers
  • DescriptionSingle Ended Mini-Din Cable, 8-Pin Conductor, 6' length