Alicat Vacuum and Pressure Gauges and Controllers
Digital display provides precise measurement and control
  • Use absolute units to measure and control vacuum
  • Accurate gauges and controllers offer digital communications capabilities
  • Simple process connection and small footprint allows for easy system integration
  • Units read PSI as standard. If you would like a different unit of measure, please contact our technical support for a custom configuration
Digital devices use multiple configurations to perform and solve pressure needs in all types of applications. From controlling low vacuum, measuring pressures up to 200 psi, or performing as a back pressure controller, these pressure meters and controllers can handle most process applications. Eliminate tedious and error-prone manual pressure meters and controllers by using the precise input and output signals with simple analog or digital signals. These signals can be used to send data to a recorder, PLC, or other pressure and flow products. The easy-to-read, 4-digit LCD display and integrated keypad around the display is all that is required to program the units for service.
226,000.00 - ₹475,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Accuracy Wetted Materials Description
Availability Pricing
Alicat Digital Pressure Gauge, 0 to 5 psig, 316L Stainless Steel and FFKM Wetted Material 4100749 Mfr # PS-5PSIG-D / 5P
COMPARE
±0.25% full-scale 316L stainless steel; FFKM standard, FKM or EPDM as required Digital Pressure Gauge, 0 to 5 psig, 316L Stainless Steel and FFKM Wetted Material
₹387,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 100 psia, for Downstream Control 6100043 Mfr # PC-100PSIA-D\5P 5INDS
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 100 psia, for Downstream Control
₹359,000.00INR / Each
Alicat Digital Pressure Gauge, 0 to 100 psig 6802354 Mfr # P-100PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Gauge, 0 to 100 psig
₹226,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 1 psig 6802653 Mfr # PC-1PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 1 psig
₹359,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 5 psig 6802654 Mfr # PC-5PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 5 psig
₹359,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 100 psig 6802658 Mfr # PC-100PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 100 psig
₹359,000.00INR / Each
Alicat Digital Pressure Gauge, 0 to 1 psig 6802738 Mfr # P-1PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Gauge, 0 to 1 psig
₹226,000.00INR / Each
Alicat Digital Pressure Gauge, 0 to 5 psig 6802740 Mfr # P-5PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Gauge, 0 to 5 psig
₹226,000.00INR / Each
Alicat Digital Pressure Gauge, 0 to 15 psig 6802742 Mfr # P-15PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Gauge, 0 to 15 psig
₹226,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 15 psig 6802744 Mfr # PC-15PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 15 psig
₹359,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 30 psig 6802746 Mfr # PC-30PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 30 psig
₹359,000.00INR / Each
Alicat PCD-15PSIG-D / 5P, 5IN Digital Pressure Controller, Dual-Valve, 0 to 15 psig 6802762 Mfr # PCD-15PSIG-D / 5P 5IN
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, Dual-Valve, 0 to 15 psig
₹475,000.00INR / Each
Alicat Digital Pressure Controller, Dual-Valve, 0 to 100 psig 6802766 Mfr # PCD-100PSIG-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, Dual-Valve, 0 to 100 psig
₹475,000.00INR / Each
Alicat Digital Pressure Controller,  0 to 30 psia 6802778 Mfr # PC-30PSIA-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 30 psia
₹359,000.00INR / Each
Alicat Digital Pressure Controller, 0 to 100 psia 6802780 Mfr # PC-100PSIA-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, 0 to 100 psia
₹359,000.00INR / Each
Alicat Digital Pressure Controller, Dual-Valve, 0 to 100 psia 6802796 Mfr # PCD-100PSIA-D
COMPARE
±0.25% full-scale 302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon Digital Pressure Controller, Dual-Valve, 0 to 100 psia
₹475,000.00INR / Each
Showing 1- 16 of 16