Alicat Digital Pressure Controller, 0 to 100 psia, for Downstream Control
Representative image only.
Alicat Digital Pressure Controller, 0 to 100 psia, for Downstream Control
Alicat | Mfr: PC-100PSIA-D\5P 5INDS| Item: 6100043
Digital display provides precise measurement and control
₹359,000.00INR / Each
₹359,000.00INR / Each
Buy It Together
Current Item
Alicat Digital Pressure Controller, 0 to 100 psia, for Downstream Control
Alicat Digital Pressure Controller, 0 to 100 psia, for Downstream Control
6100043
₹359,000.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 1 SLPM
1530881
₹169,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 200 SCCM
1530902
₹222,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 2 SLPM
1530905
₹229,000.00INR / Each
+
Alicat Power Adapter, 24 VDC, 1 Amp (Worldwide)
1530939
₹4,800.00INR / Each
=
Total Price 983800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Use absolute units to measure and control vacuum
  • Accurate gauges and controllers offer digital communications capabilities
  • Simple process connection and small footprint allows for easy system integration
  • Units read PSI as standard. If you would like a different unit of measure, please contact our technical support for a custom configuration
More About this Item
Digital devices use multiple configurations to perform and solve pressure needs in all types of applications. From controlling low vacuum, measuring pressures up to 200 psi, or performing as a back pressure controller, these pressure meters and controllers can handle most process applications. Eliminate tedious and error-prone manual pressure meters and controllers by using the precise input and output signals with simple analog or digital signals. These signals can be used to send data to a recorder, PLC, or other pressure and flow products. The easy-to-read, 4-digit LCD display and integrated keypad around the display is all that is required to program the units for service.
Single-Valve Controllers allow you to control absolute, gauge, back, and vacuum pressures. Customization is as simple as valve placement or pressure range selection. The speed of response and the stability of the unit offer many advantages over more common mechanical controllers in in-line flow-through applications.
Specifications
  • Accuracy±0.25% full-scale
  • Min Pressure (psia)0
  • Max Pressure (psia)100
  • Min Temperature (° F)14
  • Max Temperature (° F)122
  • Min Temperature (° C)-10
  • Max Temperature (DegreeC)50
  • Wetted Materials302, 303, and 304 stainless steel; FKM, alumina ceramic, glass, gold, heat-cured epoxy, heat-cured silicone rubber, polyamide, silicon
  • Process Connection1/8" NPT(F)
  • Unit Of Measurementpsig, mm Hg, " Hg, " WC, mbar, Atm, Torr, kPa
  • Display4-digit LCD, 7/16"H (1.1 cm)
  • Media CompatibilityDry, noncorrosive gases
  • DescriptionDigital Pressure Controller, 0 to 100 psia, for Downstream Control