Airfiltronix 200A HS3000A2240V Portable Fume Extractor, Double Hose; 240 VAC

Representative image only.

Airfiltronix 200A HS3000A2240V Portable Fume Extractor, Double Hose; 240 VAC

Airfiltronix | Mfr: 200A HS3000A2240V| Item: 3370546

Inexpensive solution for capturing hazardous fumes and particles

₹477,000.00INR / Each

₹477,000.00INR / Each

Buy It Together

Current Item

Airfiltronix 200A HS3000A2240V Portable Fume Extractor, Double Hose; 240 VAC
Airfiltronix 200A HS3000A2240V Portable Fume Extractor, Double Hose; 240 VAC
3370546

₹477,000.00INR / Each
+ + +
Airfiltronix FR-5 Activated Charcoal Filter, 0.5" Deep; 5/Box
3370532

₹121,000.00INR / Box of 5
+ =
Total Price 1331000.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Two 5” diameter 5-ft hoses and two tapered collector scoops.

Key Features

  • Ideal for use in work spaces too small for conventional fume hoods
  • Can be used on tabletop or wall mounted
  • Flexible 5 ft hose can easily be placed at the source
  • Accommodates a variety of filters to meet most applications
  • 1 year warranty

More About this Item

The portable fume/particle extractors are ideal for capturing hazardous fumes and particles in work areas too small for conventional fume hoods. They can be placed on tabletops or wall mounted. The extractors are available with either one or two intake scoops, each with a 5 ft heavy duty chemical resistant PVC flexible hose allowing you to easily place them at the source to capture the contaminant. A variable speed control knob operates the powerful, yet quiet 530 cfm blower.
The extractors can accommodate a variety of filters such as charcoal, four stage dacron, aluminum, and HEPA, to capture a broad range of chemical fumes, odors, and particles. Designed to improve safety and hygiene in light industrial settings, the extractors can be used in laboratory settings where a fume source can be isolated.

1 Year Warranty
Specifications
  • Length (")12 1/2
  • Width (InchShort)23 1/2
  • Height (InchShort)16
  • Exterior Width (in)23.5
  • Exterior Height (in)16
  • Exterior Depth (in)12.5
  • Power (VAC)240
  • Power (Hz)50/60
  • Exhaust Volume (CFM)273
  • DescriptionPortable Fume Extractor, Double Hose; 240 V
  • Warranty1 Year
Reviews
Q&A