Airfiltronix Portable Fume/Particle Extractors

Inexpensive solution for capturing hazardous fumes and particles
  • Ideal for use in work spaces too small for conventional fume hoods
  • Can be used on tabletop or wall mounted
  • Flexible 5 ft hose can easily be placed at the source
  • Accommodates a variety of filters to meet most applications
The portable fume/particle extractors are ideal for capturing hazardous fumes and particles in work areas too small for conventional fume hoods. They can be placed on tabletops or wall mounted. The extractors are available with either one or two intake scoops, each with a 5 ft heavy duty chemical resistant PVC flexible hose allowing you to easily place them at the source to capture the contaminant. A variable speed control knob operates the powerful, yet quiet 530 cfm blower.
The extractors can accommodate a variety of filters such as charcoal, four stage dacron, aluminum, and HEPA, to capture a broad range of chemical fumes, odors, and particles. Designed to improve safety and hygiene in light industrial settings, the extractors can be used in laboratory settings where a fume source can be isolated.
385,000.00 - ₹477,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Exhaust Volume (CFM) Exterior Height (in) Description
Availability Pricing
Airfiltronix 200A HS3000A1 Portable Fume Extractor, Single Hose; 115 VAC 3370543 Mfr # 200A HS3000A1
COMPARE
173 16 Portable Fume Extractor, Single Hose; 115 V
₹385,000.00INR / Each
Airfiltronix 200A HS3000A1240V Portable Fume Extractor, Single Hose; 240 VAC 3370544 Mfr # 200A HS3000A1240V
COMPARE
173 16 Portable Fume Extractor, Single Hose; 240 V
₹416,000.00INR / Each
Airfiltronix 200A HS3000A2 Portable Fume Extractor, Double Hose; 115 VAC 3370545 Mfr # 200A HS3000A2
COMPARE
273 16 Portable Fume Extractor, Double Hose; 115 V
₹445,000.00INR / Each
Airfiltronix 200A HS3000A2240V Portable Fume Extractor, Double Hose; 240 VAC 3370546 Mfr # 200A HS3000A2240V
COMPARE
273 16 Portable Fume Extractor, Double Hose; 240 V
₹477,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat