Airfiltronix FR-4 Activated Charcoal Filter, 2" Deep; 3/Box
Representative image only.
Airfiltronix FR-4 Activated Charcoal Filter, 2" Deep; 3/Box
Airfiltronix | Mfr: FR-4| Item: 4401160
A filter for every application
₹139,000.00INR / Box of 3
₹139,000.00INR / Box of 3
Buy It Together
Current Item
Airfiltronix FR-4 Activated Charcoal Filter, 2" Deep; 3/Box
Airfiltronix FR-4 Activated Charcoal Filter, 2" Deep; 3/Box
4401160
₹139,000.00INR / Box of 3
+ + + =
Total Price 1514000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure optimal performance of your ductless fume hood
  • Adsorbs most commonly found organic vapors and odors
  • Good for most organic solvent fumes
More About this Item
Choose from a wide range of filters to remove pollutants. These are compatible with the Aitfiltronix range of ductless fume hoods.
Specifications
  • Compatible WithAirfiltronix ductless fume hoods 33705-16 to -22, ductless fume and particulate removal workstations 33705-02 to -08, portable fume/particle extractors 33705-43 to -46, and balance enclosure 44011-00
  • Length (InchShort)17
  • Width (InchShort)10
  • Depth (InchShort)2
  • TypeActivated Charcoal Filter, 2" Deep
  • DescriptionActivated Charcoal Filter, 2" Deep; 3/Box