Airfiltronix Mini-Ductless Fume Hoods
Customize your hood with a range of filter options
  • Remove vapors and contaminates with many available filters
  • Easily view and access the workspace the clear shield
  • Quiet efficient operation powered by 530 cfm blower
These ductless fume hoods prevent airborne contaminates from escaping into the work environment, ensuring the safety of workers. Contaminated air is drawn through a variety of filters to remove pollutants. Filters are available for both vapors and particulates and they can be combined for a wide range of applications (filters sold separately). The blower and filter assembly is located on top of the enclosure to save valuable bench space. All hoods include a high-efficiency blower with backward curved impeller which moves up to 530 cfm in free air at a noise level of less than 68 dB.
The mini ductless fume hood is ideal where bench space is limited. The blower is situated on top of the enclosure and features a handle for portability. The enclosure is constructed of durable clear acrylic, making it suitable for full visibility within the hood.
507,000.00 - ₹539,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Width (in) Power (VAC) Description
Availability Pricing
Airfiltronix 200A ES7000 Mini Ductless Fume Hood, 16" W x 34" H; 120 VAC 3370541 Mfr # 200A ES7000
COMPARE
16 120 Mini Ductless Fume Hood, 16" W x 34" H; 120 VAC
₹507,000.00INR / Each
Airfiltronix 200A ES7000240V Mini Ductless Fume Hood, 16" W x 34" H; 240 VAC 3370542 Mfr # 200A ES7000 240V
COMPARE
16 240 Mini Ductless Fume Hood, 16" W x 34" H; 240 VAC
₹539,000.00INR / Each